MilliporeSigma
  • T-2 toxin-induced DNA damage in mouse livers: the effect of pretreatment with coenzyme Q10 and alpha-tocopherol.
T-2 toxin-induced DNA damage in mouse livers: the effect of pretreatment with coenzyme Q10 and alpha-tocopherol.
Molecular aspects of medicine (1997-01-01)
F Atroshi, A Rizzo, I Biese, P Veijalainen, E Antila, T Westermarck
ABSTRACT
Active oxygen species are reported to cause organ damage. This study was therefore designed to determine whether oxidative stress contributed to the initiation or progression of hepatic DNA damage produced by T-2 toxin. The aim of the study was also to investigate the behaviour of the antioxidants coenzyme Q10 (CoQ10), and alpha-tocopherol (vitamin E) against DNA damage in the livers of mice fed T-2 toxin. Treatment of fasted mice with a single dose of T-2 toxin (1.8 or 2.8 mg/kg body weight) by oral gavage led to 76% hepatic DNA fragmentation. T-2 toxin also decreased hepatic glutathione (GSH) levels markedly. Pretreatment with CoQ10 (6 mg/kg) together with alpha tocopherol (6 mg/kg) decreased DNA damage. The CoQ10 and vitamin E showed some protection against toxic cell death and glutathione depletion caused by T-2 toxin. Oxidative damage caused by T-2 toxin may be one of the underlying mechanisms for T-2 toxin-induced cell injury and DNA damage, which eventually lead to tumourigenesis.
Sigma-Aldrich
T-2 Toxin, from Fusarium sp., powder, ≥98% (HPLC)