MilliporeSigma
  • Biocide susceptibility testing of bacteria: Development of a broth microdilution method.
Biocide susceptibility testing of bacteria: Development of a broth microdilution method.
Veterinary microbiology (2020-08-23)
Angela R Schug, Alexander Bartel, Anissa D Scholtzek, Marita Meurer, Julian Brombach, Vivian Hensel, Séamus Fanning, Stefan Schwarz, Andrea T Feßler
ABSTRACT
Biocide susceptibility testing (BST) of bacteria lacks standardised methods. Based on a recently established broth macrodilution BST method, a broth microdilution method for BST was developed. To establish the respective protocol, four reference strains Staphylococcus aureus ATCC® 6538, Enterococcus hirae ATCC® 10541, Escherichia coli ATCC® 10536 and Pseudomonas aeruginosa ATCC® 15442 were investigated for their minimal inhibitory concentrations (MICs) towards quaternary ammonium compounds (benzalkonium chloride), cationic compounds (chlorhexidine), aldehydes (glutardialdehyde) and alcohols (isopropanol) using tryptic soy broth. All combinations of (i) inoculum preparation according to the German Veterinary Medical Society (DVG) or the Clinical and Laboratory Standards Institute (CLSI) with some modifications, (ii) use of 1st subculture (SC) and 2nd SC, (iii) direct colony suspension (DCS) with/without glass beads, and (iv) incubation at 37 °C for 24 h, 48 h, and 72 h were compared using seven independent replications. Overall, the reproducibility was high for all abovementioned strain/biocide/parameter combinations. In total, 86.9 % - 100 % of the results were within ± one dilution step of the mode value. The proposed method for a standardised BST protocol comprises (i) two different inoculum densities, (ii) the use of a fresh overnight culture (1st SC or 2nd SC), (iii) the preparation of the inoculum suspension by either of the two methods using DCS with or without glass beads, and (iv) the incubation at 37 °C for 24 h. This broth microdilution method will help to harmonize BST of bacterial pathogens in routine diagnostics.
Sigma-Aldrich
Chlorhexidine diacetate salt hydrate, bis(biguanide) antimicrobial