MilliporeSigma
  • A distinct metabolic response characterizes sensitivity to EZH2 inhibition in multiple myeloma.
A distinct metabolic response characterizes sensitivity to EZH2 inhibition in multiple myeloma.
Cell death & disease (2021-02-14)
Patrick Nylund, Alba Atienza Párraga, Jakob Haglöf, Elke De Bruyne, Eline Menu, Berta Garrido-Zabala, Anqi Ma, Jian Jin, Fredrik Öberg, Karin Vanderkerken, Antonia Kalushkova, Helena Jernberg-Wiklund
ABSTRACT
Multiple myeloma (MM) is a heterogeneous haematological disease that remains clinically challenging. Increased activity of the epigenetic silencer EZH2 is a common feature in patients with poor prognosis. Previous findings have demonstrated that metabolic profiles can be sensitive markers for response to treatment in cancer. While EZH2 inhibition (EZH2i) has proven efficient in inducing cell death in a number of human MM cell lines, we hereby identified a subset of cell lines that despite a global loss of H3K27me3, remains viable after EZH2i. By coupling liquid chromatography-mass spectrometry with gene and miRNA expression profiling, we found that sensitivity to EZH2i correlated with distinct metabolic signatures resulting from a dysregulation of genes involved in methionine cycling. Specifically, EZH2i resulted in a miRNA-mediated downregulation of methionine cycling-associated genes in responsive cells. This induced metabolite accumulation and DNA damage, leading to G2 arrest and apoptosis. Altogether, we unveiled that sensitivity to EZH2i in human MM cell lines is associated with a specific metabolic and gene expression profile post-treatment.
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, from mouse
Sigma-Aldrich
ChIPAb+ EZH2, clone AC22 - ChIP Validated Antibody and Primer Set, clone AC22, from mouse
Millipore
RIPA Lysis Buffer, 10X, 100 mL RIPA Lysis Buffer, 10X for Immunoprecipitation & Western Blotting.
Sigma-Aldrich
GSK343, ≥98% (HPLC)