MilliporeSigma
  • Aβ-accelerated neurodegeneration caused by Alzheimer's-associated ACE variant R1279Q is rescued by angiotensin system inhibition in mice.
Aβ-accelerated neurodegeneration caused by Alzheimer's-associated ACE variant R1279Q is rescued by angiotensin system inhibition in mice.
Science translational medicine (2020-10-02)
Leah K Cuddy, Dmitry Prokopenko, Eric P Cunningham, Ross Brimberry, Peter Song, Rory Kirchner, Brad A Chapman, Oliver Hofmann, Winston Hide, Daniele Procissi, Taleen Hanania, Steven C Leiser, Rudolph E Tanzi, Robert Vassar
ABSTRACT
Recent genome-wide association studies identified the angiotensin-converting enzyme gene (ACE) as an Alzheimer's disease (AD) risk locus. However, the pathogenic mechanism by which ACE causes AD is unknown. Using whole-genome sequencing, we identified rare ACE coding variants in AD families and investigated one, ACE1 R1279Q, in knockin (KI) mice. Similar to AD, ACE1 was increased in neurons, but not microglia or astrocytes, of KI brains, which became elevated further with age. Angiotensin II (angII) and angII receptor AT1R signaling were also increased in KI brains. Autosomal dominant neurodegeneration and neuroinflammation occurred with aging in KI hippocampus, which were absent in the cortex and cerebellum. Female KI mice exhibited greater hippocampal electroencephalograph disruption and memory impairment compared to males. ACE variant effects were more pronounced in female KI mice, suggesting a mechanism for higher AD risk in women. Hippocampal neurodegeneration was completely rescued by treatment with brain-penetrant drugs that inhibit ACE1 and AT1R. Although ACE variant-induced neurodegeneration did not depend on β-amyloid (Aβ) pathology, amyloidosis in 5XFAD mice crossed to KI mice accelerated neurodegeneration and neuroinflammation, whereas Aβ deposition was unchanged. KI mice had normal blood pressure and cerebrovascular functions. Our findings strongly suggest that increased ACE1/angII signaling causes aging-dependent, Aβ-accelerated selective hippocampal neuron vulnerability and female susceptibility, hallmarks of AD that have hitherto been enigmatic. We conclude that repurposed brain-penetrant ACE inhibitors and AT1R blockers may protect against AD.
Sigma-Aldrich
Mouse ACE  ELISA Kit, for serum, plasma and cell culture supernatant