MilliporeSigma
  • LncRNA PSMA3-AS1 promotes colorectal cancer cell migration and invasion via regulating miR-4429.
LncRNA PSMA3-AS1 promotes colorectal cancer cell migration and invasion via regulating miR-4429.
European review for medical and pharmacological sciences (2020-12-05)
P Peng, Y Wang, B-L Wang, Y-H Song, Y Fang, H Ji, C-N Huangfu, K-M Wang, Q Zheng
ABSTRACT
Many studies have revealed that long non-coding RNAs (lncRNAs) are related to various cancers, including colorectal cancer (CRC). This study aims to explore the biological function of lncRNA PSMA3-AS1 in CRC progression. The expression levels of PSMA3-AS1 and miR-4429 were assessed by RT-qPCR. CRC progression was explored by cell viability, migration, and invasion using CCK-8 and transwell assays. The interaction between PSMA3-AS1 and miR-4429 was verified by bioinformatics analysis, Dual-Luciferase assay, and RIP assay. It was found that PSMA3-AS1 expression was increased and miR-4429 expression was decreased in CRC tissues and cells. In addition, PSMA3-AS1 interference markedly hindered the proliferation, migration, and invasion of CRC cells. MiR-4429 was a direct target of PSMA3-AS1, and the knockdown of PSMA3-AS1 significantly suppressed miR-4429 expression. The depletion of PSMA3-AS1 inhibited CRC progression, which was neutralized by miR-4429 inhibitor. PSMA3-AS1 accelerated CRC progression by regulating miR-4429 expression, which could be used as a potential therapeutic target for CRC patients.
Sigma-Aldrich
MISSION® esiRNA, targeting human PSMA3