MilliporeSigma
  • Comparison of overfed Xupu and Landes geese in performance, fatty acid composition, enzymes and gene expression related to lipid metabolism.
Comparison of overfed Xupu and Landes geese in performance, fatty acid composition, enzymes and gene expression related to lipid metabolism.
Asian-Australasian journal of animal sciences (2020-02-15)
Xu Liu, Peng Li, Changqing He, Xiangyong Qu, Songchang Guo
ABSTRACT
The aim of this study was to compare overfeeding performance, fatty acid composition, blood chemistry, enzymes and genes expression overfed Xupu and Landes geese. Sixty male Xupu geese (80 d) and Landes geese (80 d) were selected. After a period of one-week of pre-overfeeding, Xupu and Landes geese were overfed three meals of 550 and 350 g/d, respectively, of a high-carbohydrate diet in the first week of the overfeeding period. The next week, geese were given four meals of 1,200 and 850 g/d, respectively, over 8 to 14 d. Finally, geese were given five meals of 1,600 and 1,350 g/d, respectively, for the last two weeks. After overfeeding for 28 d: Compared with Landes geese, Xupu geese liver weight and liver-to-body weight ratio decreased (p<0.05), while final weight, slaughter weight, total weight gain, abdominal fat weight, and feed-to-liver weight ratio increased (p<0.05). The levels of elaidic acid (C18:1t9), oleic acid (C18:1n-9), eicosenoic acid, and arachidonic acid in the liver of Xupu geese significantly increased (p<0.05), and the levels of myristic acid and stearic acid significantly decreased (p<0.05), while methyleicosanoate acid significantly increased (p<0.05). Xupu geese had higher plasma concentrations of triglyceride and very low density lipoprotein cholesterol (p<0.05), and decreased activities of alanine aminotransferase, aspartate aminotransferase, and lipase (LPS) (p<0.05). Landes geese had higher LPS activity (p<0.05), but lower cholinesterase activity (p<0.05) when compared with Xupu geese. The mRNA expression levels of fatty acid dehydrogenase (FADS) gene, elongase of longchain fatty acid 1 (ELOVL1) gene, ELOVL5, and acyl-Co A: cholesterol acyltransferase 2 (ACAT2) gene were significantly upregulated (p<0.05) in Landes goose when compared with Xupu geese. This study demonstrates that the liver production performance of Landes geese was better than that of Xupu geese to some extent, which may be closely related to LPS activity, as well as the expression of FADS, ELOVL1, ELOVL5, and ACAT2.
Product Description
Supelco
F.A.M.E. Mix, C4-C24, certified reference material,  wt. % (varied)