MilliporeSigma
  • Growth of Lactic Acid Bacteria on Gold-Influence of Surface Roughness and Chemical Composition.
Growth of Lactic Acid Bacteria on Gold-Influence of Surface Roughness and Chemical Composition.
Nanomaterials (Basel, Switzerland) (2020-12-17)
Joanna Grudzień, Magdalena Jarosz, Kamil Kamiński, Mirosława Kobasa, Karol Wolski, Marcin Kozieł, Marcin Pisarek, Grzegorz D Sulka
ABSTRACT
The main focus of this work was to establish a correlation between surface topography and chemistry and surface colonization by lactic acid bacteria. For this reason, we chose gold substrates with different surface architectures (i.e., smooth and nanorough) that were characterized by atomic force microscopy (AFM), electron scanning microscopy (SEM), and X-ray diffractometry (XRD). Moreover, to enhance biocompatibility, we modified gold substrates with polymeric monolayers, namely cationic dextran derivatives with different molar masses. The presence of those layers was confirmed by AFM, infrared spectroscopy (IR), and X-ray photoelectron spectroscopy (XPS). In order to determine the adhesion abilities of non-modified and modified gold surfaces, we tested three lactic acid bacteria (LAB) strains (i.e., Lactobacillus rhamnosus GG, Lactobacillus acidophilus, and Lactobacillus plantarum 299v). We have shown that surface roughness influences the surface colonization of bacteria, and the most significant impact on the growth was observed for the Lactobacillus rhamnosus GG strain. What is more, covering the gold surface with a molecular polymeric film by using the layer-by-layer (LbL) method allows additional changes in the bacterial growth, independently on the used strain. The well-being of the bacteria cells on tested surfaces was confirmed by using selective staining and fluorescence microscopy. Finally, we have determined the bacterial metabolic activity by measuring the amount of produced lactic acid regarding the growth conditions. The obtained results proved that the adhesion of bacteria to the metallic surface depends on the chemistry and topography of the surface, as well as the specific bacteria strain.
Sigma-Aldrich
Glycidyltrimethylammonium chloride, technical, ≥90% (calc. based on dry substance, AT)