MilliporeSigma
Targeting phosphoglycerate dehydrogenase in multiple myeloma.
Experimental hematology & oncology (2021-01-06)
Samah Elsaadi, Ida Steiro, Pegah Abdollahi, Esten N Vandsemb, Rui Yang, Tobias S Slørdahl, Torstein Baade Rø, Eline Menu, Anne-Marit Sponaas, Magne Børset
ABSTRACT
Multiple myeloma (MM) is a hematological malignancy characterized by the clonal expansion of plasma cells in the bone marrow. To date, this disease is still incurable and novel therapeutic approaches are required. Phosphoglycerate dehydrogenase (PHGDH) is the first and rate-limiting enzyme in the de novo serine synthesis pathway, and it has been attributed to bortezomib-resistance in MM. Two different PHGDH inhibitors, CBR5884 and NCT-503, were tested against human myeloma cell lines, primary MM cells from patients, and peripheral blood mononuclear cells isolated from healthy donors. The PHGDH inhibitors were then tested in combination with proteasome inhibitors in different MM cell lines, including proteasome-resistant cell lines. Furthermore, we confirmed the effects of PHGDH inhibition through knocking down PHGDH and the effect of NCT-503 in vivo in the 5T33MM mouse model. All the tested myeloma cell lines expressed PHGDH and were sensitive to doses of NCT-503 that were tolerated by peripheral blood mononuclear cells isolated from healthy donors. Upon testing bortezomib in combination with NCT-503, we noticed a clear synergy in several HMCLs. The sensitivity to bortezomib also increased after PHGDH knockdown, mimicking the effect of NCT-503 treatment. Interestingly, targeting PHGDH reduced the intracellular redox capacity of the cells. Furthermore, combination treatment with NCT-503 and bortezomib exhibited a therapeutic advantage in vivo. Our study shows the therapeutic potential of targeting PHGDH in MM, and suggest it as a way to overcome the resistance to proteasome inhibitors.
Sigma-Aldrich
NCT-503, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human PHGDH
Sigma-Aldrich
CBR-5884, ≥98% (HPLC)