MilliporeSigma
  • The effect of α-terpineol enantiomers on biomarkers of rats fed a high-fat diet.
The effect of α-terpineol enantiomers on biomarkers of rats fed a high-fat diet.
Heliyon (2020-05-10)
Gardênia Martins de Sousa, Cinthia Baú Betim Cazarin, Mário Roberto Maróstica Junior, Celina de Almeida Lamas, Valéria Helena Alves Cagnon Quitete, Glaucia Maria Pastore, Juliano Lemos Bicas
ABSTRACT
Alpha-terpineol is a monoterpenoid found in many essential oils, being widely used in food and household products. In vitro antioxidant and anti-inflammatory activities have already been associated with this alcohol; therefore, this study aimed to check if these properties were also present in vivo, counteracting the oxidant and inflammatory effects of a high-fat diet, as well as if there were differences in the biological activities among the two α-terpineol enantiomers. Thus, this work evaluated the effect of supplementation of α-terpineol enantiomers (at 25, 50 and 100 mg/kg of diet) on biological parameters of diet-induced obese Sprague-Dawley rats. In general, α-terpineol improved the nutritional parameters of rats fed a high-fat diet. The intake of α-terpineol at concentrations ≥50 mg/kg was able to reestablish the insulin sensibility and reduced (p < 0.05) serum levels of pro-inflammatory cytokines TNF-α and IL-1β, when compared with the control group. The intake of R-(+)- and (-)-α-terpineol decreased the TNF-α level by approximately 1.5 and 3.4 times, respectively, when compared with the high-fat group, regardless of the concentration. Moreover, both enantiomers at 50 mg/kg decreased the levels of serum thiobarbituric acid reactive substances (TBARS) by 2.6-4.2 times, while hepatic TBARS were reduced in approximately 1.6 times, regardless of the compound and concentration tested. Further experiments are suggested to confirm the mechanisms and the security of α-terpineol in different experimental models and more extended exposure experiments.
Product Description
Supelco
(+)-α-Terpineol, analytical standard
Pricing and availability is not currently available.