MilliporeSigma
  • Heterogeneous T cell motility behaviors emerge from a coupling between speed and turning in vivo.
Heterogeneous T cell motility behaviors emerge from a coupling between speed and turning in vivo.
eLife (2020-05-20)
Elizabeth R Jerison, Stephen R Quake
ABSTRACT
T cells in vivo migrate primarily via undirected random walks, but it remains unresolved how these random walks generate an efficient search. Here, we use light sheet microscopy of T cells in the larval zebrafish as a model system to study motility across large populations of cells over hours in their native context. We show that cells do not perform Levy flight; rather, there is substantial cell-to-cell variability in speed, which persists over timespans of a few hours. This variability is amplified by a correlation between speed and directional persistence, generating a characteristic cell behavioral manifold that is preserved under a perturbation to cell speeds, and seen in Mouse T cells and Dictyostelium. Together, these effects generate a broad range of length scales over which cells explore in vivo.
Sigma-Aldrich
Rho Kinase Inhibitor III, Rockout, The Rho Kinase Inhibitor III, Rockout, also referenced under CAS 7272-84-6, controls the biological activity of Rho Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.