MilliporeSigma
  • The Clusters of Transcription Factors NFATC2, STAT5, GATA2, AP1, RUNX1 and EGR2 Binding Sites at the Induced Il13 Enhancers Mediate Il13 Gene Transcription in Response to Antigenic Stimulation.
The Clusters of Transcription Factors NFATC2, STAT5, GATA2, AP1, RUNX1 and EGR2 Binding Sites at the Induced Il13 Enhancers Mediate Il13 Gene Transcription in Response to Antigenic Stimulation.
Journal of immunology (Baltimore, Md. : 1950) (2020-11-15)
Mohammad Kamran, Jinyi Liang, Bing Liu, Yapeng Li, Junfeng Gao, Ashley Keating, Fathia Mohamed, Shaodong Dai, Richard Reinhardt, Yang Jiong, Zhongdao Wu, Hua Huang
ABSTRACT
IL-13 plays a critical role in mediating many biological processes responsible for allergic inflammation. Mast cells express Il13 mRNA and produce IL-13 protein in response to antigenic stimulation. Enhancers are essential in promoting gene transcription and are thought to activate transcription by delivering essential accessory cofactors to the promoter to potentiate gene transcription. However, enhancers mediating Il13 have not been identified. Furthermore, which Il13 enhancers detect signals triggered by antigenic stimulation have not yet been defined. In this study, we identified potential mouse Il13 enhancers using histone modification monomethylation at lysine residue 4 on histone 3 (H3K4me1) chromatin immunoprecipitation sequencing and acetylation at lysine residue 27 on histone 3 (H3K27ac) chromatin immunoprecipitation sequencing. We used Omni-assay for transposase-accessible chromatin sequencing to determine which accessible regions within the potential Il13 enhancers that responded to IgE receptor crosslinking. We also demonstrated that the transcription factor cluster consisting of the NFATC2, STAT5, GATA2, AP1, and RUNX1 binding sites at the proximal Il13 enhancer and the transcription factor cluster consisting of the EGR2 binding site at the distal Il13 E+6.5 enhancer are critical in sensing the signals triggered by antigenic stimulation. Those enhancers, which are responsive to antigenic stimulation and are constitutively active, cooperate to generate greater transcriptional outputs. Our study reveals a novel mechanism underlying how antigenic stimulation induces robust Il13 mRNA expression in mouse mast cells.
Sigma-Aldrich
Protein A Agarose/Salmon Sperm DNA, 2.5 mL, for use in chromatin immunoprecipitations (ChIP assays)