MilliporeSigma
  • C9orf72 arginine-rich dipeptide repeats inhibit UPF1-mediated RNA decay via translational repression.
C9orf72 arginine-rich dipeptide repeats inhibit UPF1-mediated RNA decay via translational repression.
Nature communications (2020-07-06)
Yu Sun, Aziz Eshov, Jeffrey Zhou, Atagun U Isiktas, Junjie U Guo
ABSTRACT
Expansion of an intronic (GGGGCC)n repeat region within the C9orf72 gene is a main cause of familial amyotrophic lateral sclerosis and frontotemporal dementia (c9ALS/FTD). A hallmark of c9ALS/FTD is the accumulation of misprocessed RNAs, which are often targets of cellular RNA surveillance. Here, we show that RNA decay mechanisms involving upstream frameshift 1 (UPF1), including nonsense-mediated decay (NMD), are inhibited in c9ALS/FTD brains and in cultured cells expressing either of two arginine-rich dipeptide repeats (R-DPRs), poly(GR) and poly(PR). Mechanistically, although R-DPRs cause the recruitment of UPF1 to stress granules, stress granule formation is independent of NMD inhibition. Instead, NMD inhibition is primarily a result from global translational repression caused by R-DPRs. Overexpression of UPF1, but none of its NMD-deficient mutants, enhanced the survival of neurons treated by R-DPRs, suggesting that R-DPRs cause neurotoxicity in part by inhibiting cellular RNA surveillance.
Sigma-Aldrich
Anti-G3BP1 Antibody, clone 14E5-G9, clone 14E5-G9, from mouse
Sigma-Aldrich
Anti-phospho-Upf1 (Ser1127) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse