MilliporeSigma
  • Beta-lactamase-mediated beta-lactam resistance in Campylobacter species: prevalence of Cj0299 (bla OXA-61) and evidence for a novel beta-Lactamase in C. jejuni.
Beta-lactamase-mediated beta-lactam resistance in Campylobacter species: prevalence of Cj0299 (bla OXA-61) and evidence for a novel beta-Lactamase in C. jejuni.
Antimicrobial agents and chemotherapy (2009-06-10)
Deborah J Griggs, Leanne Peake, Margaret M Johnson, Saba Ghori, A Mott, Laura J V Piddock
ABSTRACT
Fifty-two percent of 1,288 poultry isolates of campylobacters were ampicillin resistant, and resistance was more common among Campylobacter coli isolates (67.4%) than among Campylobacter jejuni isolates (47.5%). Production of beta-lactamase was typically associated with resistance to ampicillin, amoxicillin (amoxicilline), penicillin, and ticarcillin. Regardless of beta-lactamase production, all isolates were resistant to piperacillin (MICs >or= 256 microg/ml), and most were resistant to carbenicillin, cloxacillin, and cephalosporins. Of all ampicillin-resistant campylobacters tested, 91% (347/380) carried the bla(OXA-61) gene, and 77% (136/175) of those tested with nitrocefin produced a beta-lactamase, presumably OXA-61. The isoelectric point (pI) of OXA-61 was 8.7, and the molecular mass was 31.0 kDa. Insertional inactivation of bla(OXA-61) in C. jejuni NCTC 11168 and two ampicillin-resistant isolates resulted in increased susceptibility to ampicillin, co-amoxiclav (amoxicillin and clavulanic acid), penicillin, carbenicillin, oxacillin, and piperacillin, but the effects on MICs of cephalosporins and imipenem were negligible. Some C. jejuni isolates that lacked bla(OXA-61) produced a beta-lactamase, CjBla2, with a pI of 9.2 and molecular mass of 32.4 kDa. Mass spectrometry confirmed that the most prevalent beta-lactamase was the product of bla(OXA-61), but CjBla2 was not identified. OXA-61 is prevalent among Campylobacter spp. of veterinary origin and is similar to the beta-lactamase previously reported in human isolates. Production of OXA-61 was associated with resistance to penams but not cephalosporins. Co-amoxiclav remained active against all isolates tested.
Product Description
Millipore
Ticarcillin Supplement, suitable for microbiology
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered
Sigma-Aldrich
Carbenicillin disodium salt, 89.0-100.5% anhydrous basis
Sigma-Aldrich
Carbenicillin disodium, meets USP testing specifications
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Carbenicillin disodium salt, BioReagent, suitable for plant cell culture