MilliporeSigma
  • Neuronal modeling of alternating hemiplegia of childhood reveals transcriptional compensation and replicates a trigger-induced phenotype.
Neuronal modeling of alternating hemiplegia of childhood reveals transcriptional compensation and replicates a trigger-induced phenotype.
Neurobiology of disease (2020-04-30)
John P Snow, Grant Westlake, Lindsay K Klofas, Soyoun Jeon, Laura C Armstrong, Kathryn J Swoboda, Alfred L George, Kevin C Ess
ABSTRACT
Alternating hemiplegia of childhood (AHC) is a rare neurodevelopmental disease caused by heterozygous de novo missense mutations in the ATP1A3 gene that encodes the neuronal specific α3 subunit of the Na,K-ATPase (NKA) pump. Mechanisms underlying patient episodes including environmental triggers remain poorly understood, and there are no empirically proven treatments for AHC. In this study, we generated patient-specific induced pluripotent stem cells (iPSCs) and isogenic controls for the E815K ATP1A3 mutation that causes the most phenotypically severe form of AHC. Using an in vitro iPSC-derived cortical neuron disease model, we found elevated levels of ATP1A3 mRNA in AHC lines compared to controls, without significant perturbations in protein expression. Microelectrode array analyses demonstrated that in cortical neuronal cultures, ATP1A3+/E815K iPSC-derived neurons displayed less overall activity than neurons differentiated from isogenic mutation-corrected and unrelated control cell lines. However, induction of cellular stress by elevated temperature revealed a hyperactivity phenotype following heat stress in ATP1A3+/E815K neurons compared to control lines. Treatment with flunarizine, a drug commonly used to prevent AHC episodes, did not impact this stress-triggered phenotype. These findings support the use of iPSC-derived neuronal cultures for studying complex neurodevelopmental conditions such as AHC and provide a platform for mechanistic discovery in a human disease model.
Sigma-Aldrich
Flunarizine dihydrochloride, ≥98% (TLC)
Supelco
Poly(ethyleneimine) solution, analytical standard, 50 % (w/v) in H2O
Roche
Laminin, from mouse Engelbreth-Holm-Swarm (EHS) sarcoma