MilliporeSigma
  • TMBIM6/BI-1 contributes to cancer progression through assembly with mTORC2 and AKT activation.
TMBIM6/BI-1 contributes to cancer progression through assembly with mTORC2 and AKT activation.
Nature communications (2020-08-13)
Hyun-Kyoung Kim, Kashi Raj Bhattarai, Raghu Patil Junjappa, Jin Hee Ahn, Suvarna H Pagire, Hyun Ju Yoo, Jaeseok Han, Duckgue Lee, Kyung-Woon Kim, Hyung-Ryong Kim, Han-Jung Chae
ABSTRACT
Transmembrane B cell lymphoma 2-associated X protein inhibitor motif-containing (TMBIM) 6, a Ca2+ channel-like protein, is highly up-regulated in several cancer types. Here, we show that TMBIM6 is closely associated with survival in patients with cervical, breast, lung, and prostate cancer. TMBIM6 deletion or knockdown suppresses primary tumor growth. Further, mTORC2 activation is up-regulated by TMBIM6 and stimulates glycolysis, protein synthesis, and the expression of lipid synthesis genes and glycosylated proteins. Moreover, ER-leaky Ca2+ from TMBIM6, a unique characteristic, is shown to affect mTORC2 assembly and its association with ribosomes. In addition, we identify that the BIA compound, a potentialTMBIM6 antagonist, prevents TMBIM6 binding to mTORC2, decreases mTORC2 activity, and also regulates TMBIM6-leaky Ca2+, further suppressing tumor formation and progression in cancer xenograft models. This previously unknown signaling cascade in which mTORC2 activity is enhanced via the interaction with TMBIM6 provides potential therapeutic targets for various malignancies.
Sigma-Aldrich
Fluorinert FC-40, immiscible fluorocarbon oil
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Insulin-like Growth Factor-I human, IGF-I, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O