MilliporeSigma
  • Net Energy Net Energy Net EnergyNet EnerNet Energy of high-protein sunflower meal fed to growing pigs and effect of dietary phosphorus on measured values of NE.
Net Energy Net Energy Net EnergyNet EnerNet Energy of high-protein sunflower meal fed to growing pigs and effect of dietary phosphorus on measured values of NE.
Journal of animal science (2020-01-18)
Jong Woong Kim, Jinyoung Lee, Charles Martin Nyachoti
ABSTRACT
An experiment was carried out to determine energy values of high-protein sunflower meal (HP-SFM) and to compare the energy values of HP-SFM determined using either a phosphorus (P)-deficient basal diet or a P-adequate basal diet. Twenty-four growing barrows were randomly assigned to 1 of 4 dietary treatments with 6 replicates per treatment. Four experimental diets including 2 basal diets containing 2 levels of standardized total tract digestible P (i.e., P-deficient and P-adequate) and the other 2 diets containing 30% HP-SFM with each basal diet (i.e., HP-SFM 1 diet and HP-SFM 2 diet) were formulated to determine the energy values of HP-SFM and to compare energy values of HP-SFM determined by the difference method using 2 basal diets. Pigs were fed diets for 15 d including 10 d for adaptation and 5 d for total collections. Pigs were then moved to indirect calorimetry chambers to determine total heat production (THP) and fasting heat production (FHP). A reduced (P < 0.01) amount of nitrogen was retained in pigs fed the P-deficient basal diet compared with those fed the other diets. The THP of pigs fed the HP-SFM 1 and 2 diets was greater (P < 0.01) than those fed the P-deficient basal diet with the intermediate value for pigs fed the P-adequate basal diet. The retained energy (RE) as protein of pigs fed the P-deficient basal diet was less (P < 0.01) but RE as lipid was greater (P < 0.01) than those fed the P-adequate basal, or HP-SFM 1 and 2 diets. However, there was no difference in FHP of pigs among the dietary treatments. The NE of HP-SFM determined using the P-deficient basal diet was 2,062 kcal/kg, as-fed basis, whereas the value determined using the P-adequate basal diet was 2,151 kcal/kg. Although no differences were observed in energy values, the amount of P in basal diet might affect energy balance by modifying N utilization, thus, a diet containing adequate amount of P is a more suitable basal diet when the difference method is used for calculation of NE in a feed ingredient.
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A