MilliporeSigma
  • Dysregulation of RyR Calcium Channel Causes the Onset of Mitochondrial Retrograde Signaling.
Dysregulation of RyR Calcium Channel Causes the Onset of Mitochondrial Retrograde Signaling.
iScience (2020-08-02)
Anindya Roy Chowdhury, Satish Srinivasan, György Csordás, György Hajnóczky, Narayan G Avadhani
ABSTRACT
This study shows that multiple modes of mitochondrial stress generated by partial mtDNA depletion or cytochrome c oxidase disruption cause ryanodine receptor channel (RyR) dysregulation, which instigates the release of Ca2+ in the cytoplasm of C2C12 myoblasts and HCT116 carcinoma cells. We also observed a reciprocal downregulation of IP3R channel activity and reduced mitochondrial uptake of Ca2+. Ryanodine, an RyR antagonist, abrogated the mitochondrial stress-mediated increase in [Ca2+]c and the entire downstream signaling cascades of mitochondrial retrograde signaling. Interestingly, ryanodine also inhibited mitochondrial stress-induced invasive behavior in mtDNA-depleted C2C12 cells and HCT116 carcinoma cells. In addition, co-immunoprecipitation shows reduced FKBP12 protein binding to RyR channel proteins, suggesting the altered function of the Ca2+ channel. These results document how the endoplasmic reticulum-associated RyR channels, in combination with inhibition of the mitochondrial uniporter system, modulate cellular Ca2+ homeostasis and signaling under mitochondrial stress conditions.
Sigma-Aldrich
2′,3′-Dideoxycytidine, ≥98% (HPLC)