MilliporeSigma
  • Development of a genetic framework to improve the efficiency of bioactive delivery from blueberry.
Development of a genetic framework to improve the efficiency of bioactive delivery from blueberry.
Scientific reports (2020-10-16)
Molla F Mengist, Haley Burtch, Hawi Debelo, Marti Pottorff, Hamed Bostan, Candace Nunn, Sydney Corbin, Colin D Kay, Nahla Bassil, Kim Hummer, Mary Ann Lila, Mario G Ferruzzi, Massimo Iorizzo
ABSTRACT
In the present study, we applied a novel high-throughput in vitro gastrointestinal digestion model to phenotype bioaccessibility of phenolics in a diverse germplasm collection representing cultivated highbush blueberries. Results revealed significant (P < 0.05) differences between accessions, years, and accession by year interaction for relative and absolute bioaccessibility of flavonoids and phenolic acids. Broad sense heritability estimates revealed low to moderate inheritances of relative and absolute bioaccessibility, suggesting that besides environmental variables, genetics factors could control bioaccessibility of phenolics. Acylated anthocyanins had significantly higher relative bioaccessibility than non-acylated anthocyanins. Correlation analysis indicated that relative bioaccessibility did not show significant association with fruit quality or raw concentration of metabolites. The study also identified accessions that have high relative and absolute bioaccessibility values. Overall, combining the bioaccessibility of phenolics with genetic and genomic approaches will enable the identification of genotypes and genetic factors influencing these traits in blueberry.
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Mucin from porcine stomach, Type II
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications