MilliporeSigma
  • Hypertonic saline mediates the NLRP3/IL-1β signaling axis in microglia to alleviate ischemic blood-brain barrier permeability by downregulating astrocyte-derived VEGF in rats.
Hypertonic saline mediates the NLRP3/IL-1β signaling axis in microglia to alleviate ischemic blood-brain barrier permeability by downregulating astrocyte-derived VEGF in rats.
CNS neuroscience & therapeutics (2020-06-13)
Qiao-Sheng Wang, Hong-Guang Ding, Sheng-Long Chen, Xin-Qiang Liu, Yi-Yu Deng, Wen-Qiang Jiang, Ya Li, Lin-Qiang Huang, Yong-Li Han, Miao-Yun Wen, Mei-Qiu Wang, Hong-Ke Zeng
ABSTRACT
The aim of this study was to explore whether the antibrain edema of hypertonic saline (HS) is associated with alleviating ischemic blood-brain barrier (BBB) permeability by downregulating astrocyte-derived vascular endothelial growth factor (VEGF), which is mediated by microglia-derived NOD-like receptor protein 3 (NLRP3) inflammasome. The infarct volume and BBB permeability were detected. The protein expression level of VEGF in astrocytes in a transient focal brain ischemia model of rats was evaluated after 10% HS treatment. Changes in the NLRP3 inflammasome, IL-1β protein expression, and the interleukin-1 receptor (IL1R1)/pNF-кBp65/VEGF signaling pathway were determined in astrocytes. HS alleviated the BBB permeability, reduced the infarct volume, and downregulated the expression of VEGF in astrocytes. HS downregulates IL-1β expression by inhibiting the activation of the NLRP3 inflammasome in microglia and then downregulates VEGF expression by inhibiting the phosphorylation of NF-кBp65 mediated by IL-1β in astrocytes. HS alleviated the BBB permeability, reduced the infarct volume, and downregulated the expression of VEGF in astrocytes. HS downregulated IL-1β expression via inhibiting the activation of the NLRP3 inflammasome in microglia and then downregulated VEGF expression through inhibiting the phosphorylation of NF-кBp65 mediated by IL-1β in astrocytes.
Sigma-Aldrich
Lectin from Lycopersicon esculentum (tomato), FITC conjugate, buffered aqueous solution