MilliporeSigma
  • Multiple convergent hypothalamus-brainstem circuits drive defensive behavior.
Multiple convergent hypothalamus-brainstem circuits drive defensive behavior.
Nature neuroscience (2020-06-24)
Matthew Lovett-Barron, Ritchie Chen, Susanna Bradbury, Aaron S Andalman, Mahendra Wagle, Su Guo, Karl Deisseroth
ABSTRACT
The hypothalamus is composed of many neuropeptidergic cell populations and directs multiple survival behaviors, including defensive responses to threats. However, the relationship between the peptidergic identity of neurons and their roles in behavior remains unclear. Here, we address this issue by studying the function of multiple neuronal populations in the zebrafish hypothalamus during defensive responses to a variety of homeostatic threats. Cellular registration of large-scale neural activity imaging to multiplexed in situ gene expression revealed that neuronal populations encoding behavioral features encompass multiple overlapping sets of neuropeptidergic cell classes. Manipulations of different cell populations showed that multiple sets of peptidergic neurons play similar behavioral roles in this fast-timescale behavior through glutamate co-release and convergent output to spinal-projecting premotor neurons in the brainstem. Our findings demonstrate that homeostatic threats recruit neurons across multiple hypothalamic cell populations, which cooperatively drive robust defensive behaviors.
Sigma-Aldrich
Tubocurarine hydrochloride pentahydrate, ≥97%