MilliporeSigma
  • Heme oxygenase-1 orchestrates the immunosuppressive program of tumor-associated macrophages.
Heme oxygenase-1 orchestrates the immunosuppressive program of tumor-associated macrophages.
JCI insight (2020-05-06)
Emmanuelle Alaluf, Benoît Vokaer, Aurélie Detavernier, Abdulkader Azouz, Marion Splittgerber, Alice Carrette, Louis Boon, Frédérick Libert, Miguel Soares, Alain Le Moine, Stanislas Goriely
ABSTRACT
Tumor-associated macrophages (TAMs) contribute to the maintenance of a strong immunosuppressive environment, supporting tumor progression and resistance to treatment. To date, the mechanisms that drive acquisition of these immunosuppressive features are still poorly defined. Heme oxygenase-1 (HO-1) is the rate-limiting enzyme that catabolizes free heme. It displays important cytoprotective, antiinflammatory, and antioxidant properties. A growing body of evidence suggests that HO-1 may also promote tumor development. Herein, we show that HO-1 is highly expressed in monocytic cells in the tumor microenvironment (TME) once they differentiate into TAMs. Deletion of HO-1 in the myeloid compartment enhances the beneficial effects of a therapeutic antitumor vaccine by restoring CD8+ T cell proliferation and cytotoxicity. We further show that induction of HO-1 plays a major role in monocyte education by tumor cells by modulating their transcriptional and epigenetic programs. These results identify HO-1 as a valuable therapeutic target to reprogram the TME and synergize with current cancer therapies to facilitate antitumor response.
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Roche
DNase I, grade II, from bovine pancreas