MilliporeSigma
  • A one-step, one-tube real-time RT-PCR based assay with an automated analysis for detection of SARS-CoV-2.
A one-step, one-tube real-time RT-PCR based assay with an automated analysis for detection of SARS-CoV-2.
Heliyon (2020-07-16)
Bhasker Dharavath, Neelima Yadav, Sanket Desai, Roma Sunder, Rohit Mishra, Madhura Ketkar, Prasanna Bhanshe, Anurodh Gupta, Archana Kumari Redhu, Nikhil Patkar, Shilpee Dutt, Sudeep Gupta, Amit Dutt
ABSTRACT
Early diagnosis of SARS-CoV-2 infected patients is essential to control the dynamics of the COVID-19 pandemic. We develop a rapid and accurate one-step multiplex TaqMan probe-based real-time RT-PCR assay, along with a computational tool to systematically analyse the data. Our assay could detect to a limit of 15 copies of SARS-CoV-2 transcripts-based on experiments performed by spiking total human RNA with in vitro synthesized viral transcripts. The assay was evaluated by performing 184 validations for the SARS-CoV-2 Nucleocapsid gene and human RNase P as an internal control reference gene with dilutions ranging from 1-100 ng for human RNA on a cohort of 26 clinical samples. 5 of 26 patients were confirmed to be infected with SARS-CoV-2, while 21 tested negative, consistent with the standards. The accuracy of the assay was found to be 100% sensitive and 100% specific based on the 26 clinical samples that need to be further verified using a large number of clinical samples. In summary, we present a rapid, easy to implement real-time PCR based assay with automated analysis using a novel COVID qPCR Analyzer tool with graphical user interface (GUI) to analyze the raw qRT-PCR data in an unbiased manner at a cost of under $3 per reaction and turnaround time of less than 2h, to enable in-house SARS-CoV-2 testing across laboratories.
Sigma-Aldrich
KiCqStart® One-Step Probe RT-qPCR ReadyMix, for Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems