MilliporeSigma
  • Reactive-oxygen-species-mediated mechanism for photoinduced antibacterial and antiviral activities of Ag3PO4.
Reactive-oxygen-species-mediated mechanism for photoinduced antibacterial and antiviral activities of Ag3PO4.
Journal of analytical science and technology (2020-06-17)
Youngsik Seo, Keunchun Park, Yonggun Hong, Eun Sik Lee, Sang-Soon Kim, Yong-Tae Jung, Heonyong Park, Chian Kwon, Young-Sik Cho, Young-Duk Huh
ABSTRACT
Cubic-shaped Ag3PO4 crystals with a mean size of 1 μm were synthesized by a precipitation method from a mixed solution of AgNO3, Na2HPO4, and triethanolamine. The antibacterial activities against Escherichia coli, Listeria innocua, and Pseudomonas syringae DC3000 in both the absence and presence of Ag3PO4 under dark conditions and in the presence of Ag3PO4 under red-light (625 nm) and blue-light (460 nm) irradiation were examined. The concentrations of reactive oxygen species (ROS) were also measured in the antibacterial action of the Ag3PO4 against Escherichia coli. The photoinduced enhancement of the Ag3PO4 antibacterial activity under blue-light irradiation is explained by the formation of ROS during the antibacterial action of the Ag3PO4. Moreover, the antiviral activity of Ag3PO4 against amphotropic 10A1 murine leukemia virus enhanced under blue-light irradiation via ROS production. These results provide an insight into extended bio-applications of Ag3PO4.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)