MilliporeSigma
  • Mechanical, Electrical, and Thermal Properties of Carbon Nanotube Buckypapers/Epoxy Nanocomposites Produced by Oxidized and Epoxidized Nanotubes.
Mechanical, Electrical, and Thermal Properties of Carbon Nanotube Buckypapers/Epoxy Nanocomposites Produced by Oxidized and Epoxidized Nanotubes.
Materials (Basel, Switzerland) (2020-10-01)
George Trakakis, Georgia Tomara, Vitaliy Datsyuk, Labrini Sygellou, Asterios Bakolas, Dimitrios Tasis, John Parthenios, Christoforos Krontiras, Stavroula Georga, Costas Galiotis, Kostas Papagelis
ABSTRACT
High volume fraction carbon nanotube (CNT) composites (7.5-16% vol.) were fabricated by the impregnation of CNT buckypapers into epoxy resin. To enhance the interfacial reaction with the epoxy resin, the CNTs were modified by two different treatments, namely, an epoxidation treatment and a chemical oxidation. The chemical treatment was found to result in CNT length severance and to affect the porosity of the buckypapers, having an important impact on the physico-mechanical properties of the nanocomposites. Overall, the mechanical, electrical, and thermal properties of the impregnated buckypapers were found to be superior of the neat epoxy resin, offering an attractive combination of mechanical, electrical, and thermal properties for multifunctional composites.
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Potassium permanganate, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
3-Chloroperbenzoic acid, ≤77%
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)