MilliporeSigma
  • SCRINSHOT enables spatial mapping of cell states in tissue sections with single-cell resolution.
SCRINSHOT enables spatial mapping of cell states in tissue sections with single-cell resolution.
PLoS biology (2020-11-21)
Alexandros Sountoulidis, Andreas Liontos, Hong Phuong Nguyen, Alexandra B Firsova, Athanasios Fysikopoulos, Xiaoyan Qian, Werner Seeger, Erik Sundström, Mats Nilsson, Christos Samakovlis
ABSTRACT
Changes in cell identities and positions underlie tissue development and disease progression. Although single-cell mRNA sequencing (scRNA-Seq) methods rapidly generate extensive lists of cell states, spatially resolved single-cell mapping presents a challenging task. We developed SCRINSHOT (Single-Cell Resolution IN Situ Hybridization On Tissues), a sensitive, multiplex RNA mapping approach. Direct hybridization of padlock probes on mRNA is followed by circularization with SplintR ligase and rolling circle amplification (RCA) of the hybridized padlock probes. Sequential detection of RCA-products using fluorophore-labeled oligonucleotides profiles thousands of cells in tissue sections. We evaluated SCRINSHOT specificity and sensitivity on murine and human organs. SCRINSHOT quantification of marker gene expression shows high correlation with published scRNA-Seq data over a broad range of gene expression levels. We demonstrate the utility of SCRINSHOT by mapping the locations of abundant and rare cell types along the murine airways. The amenability, multiplexity, and quantitative qualities of SCRINSHOT facilitate single-cell mRNA profiling of cell-state alterations in tissues under a variety of native and experimental conditions.
Sigma-Aldrich
SSC Buffer 20× Concentrate, for Northern and Southern blotting, solution
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology