MilliporeSigma
  • A non-diploid DNA status is linked to poor prognosis in renal cell cancer.
A non-diploid DNA status is linked to poor prognosis in renal cell cancer.
World journal of urology (2020-05-04)
Franziska Büscheck, Christoph Fraune, Martina Kluth, Maximilian Lennartz, Ronald Simon, Claudia Hube-Magg, Christian Morlock, Silvano Barbieri, Carolin Wahl, Christian Eichelberg, Christina Möller-Koop, Doris Höflmayer, Corinna Wittmer, Waldemar Wilczak, Guido Sauter, Margit Fisch, Till Eichenauer, Michael Rink
ABSTRACT
DNA ploidy measurement has earlier been suggested as a potentially powerful prognostic tool in many cancer types, but the role in renal tumors is still unclear. To clarify its prognostic impact, we analyzed the DNA content of 1320 kidney tumors, including clear cell, papillary and chromophobe renal cell carcinoma (RCC) as well as renal oncocytoma and compared these data with clinico-pathological parameters and patient prognosis. A non-diploid DNA content was seen in 37% of 1276 analyzable renal tumors with a striking predominance in chromophobe carcinoma (74.3% of 70 cases). In clear cell carcinoma, a non-diploid DNA content was significantly linked to high-grade (ISUP, Fuhrman, Thoenes; p < 0.0001 each), advanced tumor stage (p = 0.0011), distant metastasis (p < 0.0001), shortened overall survival (p = 0.0010), and earlier recurrence (p < 0.0001). In papillary carcinoma, an aberrant DNA content was significantly linked to high Fuhrman grade (p = 0.0063), distant metastasis (p = 0.0138), shortened overall survival (p = 0.0010), and earlier recurrence (p = 0.0003). In summary, the results of our study identify a non-diploid DNA content as a predictor of an unfavorable prognosis in clear cell and papillary carcinoma.
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Propidium iodide solution