MilliporeSigma
  • An Insight into Kiwiberry Leaf Valorization: Phenolic Composition, Bioactivity and Health Benefits.
An Insight into Kiwiberry Leaf Valorization: Phenolic Composition, Bioactivity and Health Benefits.
Molecules (Basel, Switzerland) (2021-05-01)
Ana Margarida Silva, Diana Pinto, Iva Fernandes, Victor de Freitas, María de la Luz Cádiz-Gurrea, Paulo Costa, Cristina Delerue-Matos, Francisca Rodrigues
ABSTRACT
During kiwiberry production, different by-products are generated, including leaves that are removed to increase the fruit's solar exposure. The aim of this work was to extract bioactive compounds from kiwiberry leaf by employing microwave-assisted extraction (MAE). Compatible food solvents (water and ethanol) were employed. The alcoholic extract contained the highest phenolic and flavonoid contents (629.48 mg of gallic acid equivalents (GAE) per gram of plant material on dry weight (dw) (GAE/g dw) and 136.81 mg of catechin equivalents per gram of plant material on dw (CAE/g dw), respectively). Oppositely, the hydroalcoholic extract achieved the highest antioxidant activity and scavenging activity against reactive oxygen and nitrogen species (IC50 = 29.10 μg/mL for O2•-, IC50 = 1.87 μg/mL for HOCl and IC50 = 1.18 μg/mL for •NO). The phenolic profile showed the presence of caffeoylquinic acids, proanthocyanidin, and quercetin in all samples. However, caffeoylquinic acids and quercetin were detected in higher amounts in the alcoholic extract, while proanthocyanidins were prevalent in the hydroalcoholic extract. No adverse effects were observed on Caco-2 viability, while the highest concentration (1000 µg/mL) of hydroalcoholic and alcoholic extracts conducted to a decrease of HT29-MTX viability. These results highlight the MAE potentialities to extract bioactive compounds from kiwiberry leaf.
Sigma-Aldrich
Amylase Activity Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
Acetylcholinesterase Activity Assay Kit, sufficient for 100 colorimetric tests