MilliporeSigma
  • A reliable set of reference genes to normalize oxygen-dependent cytoglobin gene expression levels in melanoma.
A reliable set of reference genes to normalize oxygen-dependent cytoglobin gene expression levels in melanoma.
Scientific reports (2021-05-27)
Joey De Backer, Darko Maric, Matthias Bosman, Sylvia Dewilde, David Hoogewijs
ABSTRACT
Cytoglobin (CYGB) is a ubiquitously expressed protein with a protective role against oxidative stress, fibrosis and tumor growth, shown to be transcriptionally regulated under hypoxic conditions. Hypoxia-inducible CYGB expression is observed in several cancer cell lines and particularly in various melanoma-derived cell lines. However, reliable detection of hypoxia-inducible mRNA levels by qPCR depends on the critical choice of suitable reference genes for accurate normalization. Limited evidence exists to support selection of the commonly used reference genes in hypoxic models of melanoma. This study aimed to select the optimal reference genes to study CYGB expression levels in melanoma cell lines exposed to hypoxic conditions (0.2% O2) and to the HIF prolyl hydroxylase inhibitor roxadustat (FG-4592). The expression levels of candidate genes were assessed by qPCR and the stability of genes was evaluated using the geNorm and NormFinder algorithms. Our results display that B2M and YWHAZ represent the most optimal reference genes to reliably quantify hypoxia-inducible CYGB expression in melanoma cell lines. We further validate hypoxia-inducible CYGB expression on protein level and by using CYGB promoter-driven luciferase reporter assays in melanoma cell lines.
Product Description
Mouse IgG HRP Linked Whole Ab, Cytiva NA931-1ML
Rabbit IgG HRP Linked Whole Ab, Cytiva NA934-1ML