MilliporeSigma
Chromatin-Bound Proteome Profiling by Genome Capture.
STAR protocols (2020-10-29)
Sergi Aranda, Eva Borràs, Eduard Sabidó, Luciano Di Croce
ABSTRACT
De novo identification of chromatin interactors can reveal unexpected pathways relevant to physiology and human disease. Inspired by the DNA mediated chromatin pull-down (Dm-ChP) technology (also known as iPOND [isolation of proteins on nascent DNA]) for the proteomic characterization of nascent DNA, we have recently reported a new experimental protocol that allows for the identification of proteins on total DNA (iPOTD) for bulk chromatome profiling and de novo identification of chromatin-bound proteins. Here, we detail a step-by-step protocol to survey the cellular chromatin-bound proteome in a simple, robust, and unbiased manner. For complete details on the use and execution of this protocol, please refer to Aranda et al. (2019).
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
β-Glycerol phosphate disodium salt pentahydrate, ≥98.0% (NT)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.