MilliporeSigma
  • Synthesis and antibacterial property of quinolines with potent DNA gyrase activity.
Synthesis and antibacterial property of quinolines with potent DNA gyrase activity.
Bioorganic & medicinal chemistry (2008-12-23)
Ekambaram Ramesh, Rathna Durga R S Manian, Ragavachary Raghunathan, Shilpakala Sainath, Malathi Raghunathan
ABSTRACT
Synthesis of a series of novel tetrahydroquinoline annulated heterocycles has been accomplished by intramolecular imino and bisimino Diels-Alder reaction. These compounds were evaluated for their antibacterial activity. All the synthetic compounds, exhibited good antibacterial activity against microorganisms of which one of them 7 was found to be as active as the antibiotic ciplofloxacin and is found to have MIC value of 2.5 mg/mL against Escherichia coli.
Sigma-Aldrich
Nalidixic acid, ≥98%
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard