MilliporeSigma
  • Tumour-Secreted Hsp90α on External Surface of Exosomes Mediates Tumour - Stromal Cell Communication via Autocrine and Paracrine Mechanisms.
Tumour-Secreted Hsp90α on External Surface of Exosomes Mediates Tumour - Stromal Cell Communication via Autocrine and Paracrine Mechanisms.
Scientific reports (2019-10-24)
Xin Tang, Cheng Chang, Jiacong Guo, Vadim Lincoln, Chengyu Liang, Mei Chen, David T Woodley, Wei Li
ABSTRACT
Extracellular heat shock protein-90alpha (eHsp90α) plays an essential role in tumour invasion and metastasis. The plasma eHsp90α levels in patients with various cancers correlate with the stages of the diseases. Nonetheless, the mechanism of action by tumour-secreted eHsp90α remained unclear. Here we show that eHsp90α accounts for approximately 1% of the total cellular Hsp90α and is associated with tumour-secreted exosomes. CRISPR-cas9 knockout of Hsp90α did not affect the overall distribution and quantity of secreted exosomes, but it caused increased exosome-associated CD9 and decreased exosome-associated TSG101, Alix, and CD63. However, Hsp90α-knockout tumour cells have not only lost their own constitutive motility, but also the ability to recruit stromal cells via secreted exosomes. These defects are specifically due to the lack of eHsp90α on tumour cell-secreted exosomes. Anti-Hsp90α antibody nullified the pro-motility activity of tumour-secreted exosomes and human recombinant Hsp90α protein fully rescued the functional defects of eHsp90α-free exosomes. Finally, while current exosome biogenesis models exclusively implicate the luminal location of host cytosolic proteins inside secreted exosomes, we provide evidence for eHsp90α location on the external surface of tumour-secreted exosomes. Taken together, this study elucidates a new mechanism of action by exosome-associated eHsp90α.
Sigma-Aldrich
Anti-Hsp90α Mouse mAb (EMD-17D7), liquid, clone EMD-17D7, Calbiochem®