MilliporeSigma
  • Diaryl-dithiolanes and -isothiazoles: COX-1/COX-2 and 5-LOX-inhibitory, *OH scavenging and anti-adhesive activities.
Diaryl-dithiolanes and -isothiazoles: COX-1/COX-2 and 5-LOX-inhibitory, *OH scavenging and anti-adhesive activities.
Bioorganic & medicinal chemistry (2008-12-23)
Michael Scholz, Holger K Ulbrich, Oliver Soehnlein, Lennart Lindbom, Andreas Mattern, Gerd Dannhardt
ABSTRACT
Three series of non-steroidal anti-inflammatory drugs (NSAIDs) inhibiting the cyclooxygenase/5-lipoxygenase (COX/5-LOX) pathways as such as formation of hydroxyl radicals and adhesion were prepared: 4,5-diaryl isothiazoles, 4,5-diaryl 3H-1,2-dithiole-3-thiones and 4,5-diaryl 3H-1,2-dithiole-3-ones. The aim of the present study was to develop substances which can intervene into the inflammatory processes via different mechanisms of action as multiple target non-steroidal anti-inflammatory drugs (MTNSAIDs) with increased anti-inflammatory potential. The current lead 11a was evaluated in COX-1/2, 5-LOX and (*)OH scavenging in vitro assays and in a static adhesion assay where it proved to inhibit adhesion. Moreover, 11a treatment attenuated expression of macrophage adhesion molecule-1 (Mac-1) on extravasated polymorphonuclear leukocytes (PMNs) which indicates that the activation was reduced. The assays used are predictive for the in vivo efficacy of test compounds as shown for 11a in a peritonitis model of acute inflammation in mice. Thus, the novel 5-LOX/COX and (*)OH inhibitor 11a possesses anti-inflammatory activity that, in addition to COX/5-LOX inhibition, implicates effects on leukocyte-endothelial interactions.
Product Description
Supelco
(R)-(+)-α-Lipoic acid, analytical standard
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)