MilliporeSigma
  • Features of adult neurogenesis and neurochemical signaling in the Cherry salmon Oncorhynchus masou brain.
Features of adult neurogenesis and neurochemical signaling in the Cherry salmon Oncorhynchus masou brain.
Neural regeneration research (2013-01-05)
Evgeniya V Pushchina, Dmitry K Obukhov, Anatoly A Varaksin
ABSTRACT
We investigated the distribution of gamma aminobutyric acid, tyrosine hydroxylase and nitric oxide-producing elements in a cherry salmon Oncorhynchus masou brain at various stages of postnatal ontogenesis by immunohistochemical staining and histochemical staining. The periventricular region cells exhibited the morphology of neurons and glia including radial glia-like cells and contained several neurochemical substances. Heterogeneous populations of tyrosine hydroxylase-, gamma aminobutyric acid-immunoreactive, as well as nicotinamide adenine dinucleotide phosphate diaphorase-positive cells were observed in proliferating cell nuclear antigen-immunoreactive proliferative zones in periventricular area of diencephalon, central grey layer of dorsomedial tegmentum, medulla and spinal cord. Immunolocalization of Pax6 in the cherry salmon brain revealed a neuromeric construction of the brain at various stages of postnatal ontogenesis, and this was confirmed by tyrosine hydroxylase and gamma aminobutyric acid labeling.