MilliporeSigma
  • Carbonic anhydrase inhibitors: inhibition of the beta-class enzyme from the yeast Saccharomyces cerevisiae with sulfonamides and sulfamates.
Carbonic anhydrase inhibitors: inhibition of the beta-class enzyme from the yeast Saccharomyces cerevisiae with sulfonamides and sulfamates.
Bioorganic & medicinal chemistry (2009-01-07)
Semra Isik, Feray Kockar, Meltem Aydin, Oktay Arslan, Ozen Ozensoy Guler, Alessio Innocenti, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
The protein encoded by the Nce103 gene of Saccharomyces cerevisiae, a beta-carbonic anhydrase (CA, EC 4.2.1.1) designated as scCA, has been cloned, purified, characterized kinetically and investigated for its inhibition with a series of sulfonamides and one sulfamate. The enzyme showed high CO(2) hydrase activity, with a k(cat) of 9.4x10(5)s(-1), and k(cat)/K(M) of 9.8x10(7)M(-1)s(-1). Simple benzenesulfonamides substituted in 2-, 4- and 3,4-positions of the benzene ring with amino, alkyl, halogeno and hydroxyalkyl moieties were weak scCA inhibitors with K(I)s in the range of 0.976-18.45 microM. Better inhibition (K(I)s in the range of 154-654 nM) was observed for benzenesulfonamides incorporating aminoalkyl/carboxyalkyl moieties or halogenosulfanilamides; benzene-1,3-disulfonamides; simple heterocyclic sulfonamides and sulfanilyl-sulfonamides. The clinically used sulfonamides/sulfamate (acetazolamide, ethoxzolamide, methazolamide, dorzolamide, topiramate, celecoxib, etc.) generally showed effective scCA inhibitory activity, with K(I)s in the range of 82.6-133 nM. The best inhibitor (K(I) of 15.1 nM) was 4-(2-amino-pyrimidin-4-yl)-benzenesulfonamide. These inhibitors may be useful to better understand the physiological role of beta-CAs in yeast and some pathogenic fungi which encode orthologues of the yeast enzyme and eventually for designing novel antifungal therapies.
Sigma-Aldrich
Saccharin, ≥98%
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
4-Sulfamoylbenzoic acid, 97%
Sigma-Aldrich
(±)-Sulpiride
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
p-Toluenesulfonamide, reagent grade, 97%
Sigma-Aldrich
p-Toluenesulfonamide, ReagentPlus®, ≥99%