MilliporeSigma
  • The Myc-dependent angiogenic switch in tumors is mediated by interleukin 1beta.
The Myc-dependent angiogenic switch in tumors is mediated by interleukin 1beta.
Genes & development (2006-09-19)
Ksenya Shchors, Elena Shchors, Fanya Rostker, Elizabeth R Lawlor, Lamorna Brown-Swigart, Gerard I Evan
ABSTRACT
Although induction of blood vessel growth is acknowledged as a pivotal requirement for the evolution of macroscopic tumors, the events that trigger onset of tumor angiogenesis remain largely obscure. The pervasive Myc oncoprotein is itself a potent inducer of angiogenesis in a wide range of tissues. We have used a reversibly switchable mouse transgenic model of Myc-dependent beta-cell carcinogenesis to delineate the kinetics and causal sequence of angiogenic processes following acute Myc activation. We show that onset of endothelial cell proliferation is induced shortly after Myc-induced cell cycle entry of beta cells. Endothelial cell proliferation is not indirectly induced by local tissue hypoxia but instead via a diffusible angiogenic signal produced by Myc-expressing beta cells. This signal triggers the release of pre-existing, sequestered VEGF from the islet extracellular matrix, that then homes to the endothelial compartment where it induces endothelial cell proliferation. Myc activation in beta cells rapidly induces expression and release of the proinflammatory cytokine interleukin 1beta (IL-1beta). We show that IL-1beta is the principal effector downstream of Myc responsible for triggering rapid onset of islet angiogenesis. Together, our data delineate a complete pathway in vivo by which the highly pleiotropic Myc oncoproteins elicits coexpansion of the vascular compartment during tumorigenic progression.
Sigma-Aldrich
Fibrin In Vitro Angiogenesis Assay, The Fibrin Gel In Vitro Angiogenesis Assay Kit represents a simple model of angiogenesis in which the induction or inhibition of tube formation by exogenous signals can be easily monitored.