MilliporeSigma
  • Antioxidant and cytoprotective effects of Stachys riederi var. japonica ethanol extract on UVA‑irradiated human dermal fibroblasts.
Antioxidant and cytoprotective effects of Stachys riederi var. japonica ethanol extract on UVA‑irradiated human dermal fibroblasts.
International journal of molecular medicine (2019-01-11)
Ji Yeon Hwang, Anil Kumar Yadav, Byeong-Churl Jang, Young Chul Kim
ABSTRACT
Stachys riederi is one of the largest genera in the flowering plant family Lamiaceae. The aqueous extract of Stachys riederi var. japonica is known for its anti‑allergic effect. In the present study, the antioxidant and cytoprotective effects of Stachys riederi var. japonica ethanol extract (SREE) on ultraviolet A (UVA)‑irradiated human dermal fibroblasts (HDFs) were evaluated. At 100 µg/ml, SREE significantly inhibited production of reactive oxygen species (ROS) in UVA‑irradiated HDFs. SREE at 100 µg/ml additionally markedly interfered with the loss of mitochondrial membrane potential (ΔΨm) in these cells. In addition, SREE at 100 µg/ml attenuated UVA‑induced DNA fragmentation and caspase‑3 activation in HDFs. SREE at 100 µg/ml additionally increased mRNA and protein expressions of Bcl‑2 and decreased those of Bax and cytochrome c in UVA‑irradiated HDFs. In summary, to the best of our knowledge, these results demonstrate for the first time that SREE exhibited antioxidant and cytoprotective effects on UVA‑irradiated HDFs, which may be mediated through suppression of ROS generation, inhibition of the loss of ΔΨm and inhibition of apoptosis.
Sigma-Aldrich
Caspase 3 Colorimetric Activity Assay Kit, DEVD, Caspase-3 Colorimetric Activity Assay Kits provide a simple & convenient means for assaying the activity of caspases that recognize the sequence DEVD.
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline