MilliporeSigma
  • The Effects of Different Purifying Methods on the Chemical Properties, in Vitro Anti-Tumor and Immunomodulatory Activities of Abrus cantoniensis Polysaccharide Fractions.
The Effects of Different Purifying Methods on the Chemical Properties, in Vitro Anti-Tumor and Immunomodulatory Activities of Abrus cantoniensis Polysaccharide Fractions.
International journal of molecular sciences (2016-04-09)
Shaowei Wu, Xiong Fu, Margaret A Brennan, Charles S Brennan, Chen Chun
ABSTRACT
Abrus cantoniensis (Hance) is a popular Chinese vegetable consumed as a beverage, soup or folk medicine. To fully exploit the potential of the polysaccharide in Abrus cantoniensis, nine polysaccharide fractions of Abrus cantoniensis were isolated and purified (AP-AOH30-1, AP-AOH30-2, AP-AOH80-1, AP-AOH80-2, AP-ACl-1, AP-ACl-2, AP-ACl-3, AP-H and AP-L). Fourier-transform infrared spectroscopy (FT-IR) and gas chromatography (GC) were used to characterize these Abrus polysaccharides fractions (APF). In vitro anti-tumor and immunomodulatory activities were also investigated and compared using the rank-sum ratio (RSR) method. Results demonstrated significant differences in the structure and bioactivities among APF, which were associated to the process used for their purification. Among the APF, AP-ACl-3 yield was 613.5 mg/kg of product and consisted of rhamnose (9.8%), arabinose (8.9%), fructose (3.0%), galactose (9.9%), glucose (4.3%), galacturonic acid (3.0%) and glucuronic acid (61.1%) with a molecular weight of 4.4 × 10⁴ Da. Furthermore, AP-ACl-3 exhibited considerable bioactivities significantly preventing the migration of MCF-7 cells and stimulating lymphocyte proliferation along with nitric oxide (NO) production of peritoneal macrophages. AP-ACl-3 could be explored as a novel potential anti-tumor and immunomodulatory agent.