MilliporeSigma
  • Young to Middle-Aged Dogs with High Amyloid-β Levels in Cerebrospinal Fluid are Impaired on Learning in Standard Cognition tests.
Young to Middle-Aged Dogs with High Amyloid-β Levels in Cerebrospinal Fluid are Impaired on Learning in Standard Cognition tests.
Journal of Alzheimer's disease : JAD (2016-12-31)
Herman Borghys, Bianca Van Broeck, Deborah Dhuyvetter, Tom Jacobs, Katja de Waepenaert, Tim Erkens, Melissa Brooks, Sandy Thevarkunnel, Joseph A Araujo
ABSTRACT
Understanding differences in Alzheimer's disease biomarkers before the pathology becomes evident can contribute to an improved understanding of disease pathogenesis and treatment. A decrease in amyloid-β (Aβ)42 in cerebrospinal fluid (CSF) is suggested to be a biomarker for Aβ deposition in brain. However, the relevance of CSF Aβ levels prior to deposition is not entirely known. Dogs are similar to man with respect to amyloid-β protein precursor (AβPP)-processing, age-related amyloid plaque deposition, and cognitive dysfunction. In the current study, we evaluated the relation between CSF Aβ42 levels and cognitive performance in young to middle-aged dogs (1.5-7 years old). Additionally, CSF sAβPPα and sAβPPβ were measured to evaluate AβPP processing, and CSF cytokines were measured to determine the immune status of the brain. We identified two groups of dogs showing consistently low or high CSF Aβ42 levels. Based on prior studies, it was assumed that at this age no cerebral amyloid plaques were likely to be present. The cognitive performance was evaluated in standard cognition tests. Low or high Aβ concentrations coincided with low or high sAβPPα, sAβPPβ, and CXCL-1 levels, respectively. Dogs with high Aβ concentrations showed significant learning impairments on delayed non-match to position (DNMP), object discrimination, and reversal learning compared to dogs with low Aβ concentrations. Our data support the hypothesis that high levels of CSF Aβ in dogs coincide with lower cognitive performance prior to amyloid deposition. Further experiments are needed to investigate this link, as well as the relevance with respect to Alzheimer's disease pathology progression.
Product Description
Millipore
MILLIPLEX® Canine Cytokine/Chemokine Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.