MilliporeSigma
  • A bacterial symbiont is converted from an inedible producer of beneficial molecules into food by a single mutation in the gacA gene.
A bacterial symbiont is converted from an inedible producer of beneficial molecules into food by a single mutation in the gacA gene.
Proceedings of the National Academy of Sciences of the United States of America (2013-07-31)
Pierre Stallforth, Debra A Brock, Alexandra M Cantley, Xiangjun Tian, David C Queller, Joan E Strassmann, Jon Clardy
ABSTRACT
Stable multipartite mutualistic associations require that all partners benefit. We show that a single mutational step is sufficient to turn a symbiotic bacterium from an inedible but host-beneficial secondary metabolite producer into a host food source. The bacteria's host is a "farmer" clone of the social amoeba Dictyostelium discoideum that carries and disperses bacteria during its spore stage. Associated with the farmer are two strains of Pseudomonas fluorescens, only one of which serves as a food source. The other strain produces diffusible small molecules: pyrrolnitrin, a known antifungal agent, and a chromene that potently enhances the farmer's spore production and depresses a nonfarmer's spore production. Genome sequence and phylogenetic analyses identify a derived point mutation in the food strain that generates a premature stop codon in a global activator (gacA), encoding the response regulator of a two-component regulatory system. Generation of a knockout mutant of this regulatory gene in the nonfood bacterial strain altered its secondary metabolite profile to match that of the food strain, and also, independently, converted it into a food source. These results suggest that a single mutation in an inedible ancestral strain that served a protective role converted it to a "domesticated" food source.
Sigma-Aldrich
GenElute Bacterial Genomic DNA Kits, sufficient for 10 purifications