MilliporeSigma
  • Synthesis and evaluation of 1-(benzo[b]thiophen-2-yl)ethanone analogues as novel anti-osteoporosis agents acting on BMP-2 promotor.
Synthesis and evaluation of 1-(benzo[b]thiophen-2-yl)ethanone analogues as novel anti-osteoporosis agents acting on BMP-2 promotor.
Bioorganic & medicinal chemistry letters (2009-06-16)
Zong-ying Liu, Xiao-bo He, Zhao-yong Yang, Hua-yi Shao, Xue Li, Hui-fang Guo, Yue-qin Zhang, Shu-yi Si, Zhuo-rong Li
ABSTRACT
A novel series of 1-(benzo[b]thiophen-2-yl)ethanone analogues were prepared and evaluated for enhancing BMP-2 expression. Compounds 1-5, 7, 8, 12, 13 and 16, with upregulation rate values of 35.6%, 27.9%, 39.8%, 32.0%, 37.1%, 30.2%, 28.0%, 33.5%, 22.8% and 27.3% in vitro, respectively, at a concentration of 4muM, exhibited potent effect for enhancing BMP-2 expression. We also found that compounds 1 and 12 produced a dose-dependent increase on bone histology and histomorphometry, and effectively reduced bone defects induced by ovariectomy in an ovariectomized rat model (OVX).
Sigma-Aldrich
3-Acetylindole, 98%
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)