MilliporeSigma
  • Analysis of oxidative events induced by expanded polyglutamine huntingtin exon 1 that are differentially restored by expression of heat shock proteins or treatment with an antioxidant.
Analysis of oxidative events induced by expanded polyglutamine huntingtin exon 1 that are differentially restored by expression of heat shock proteins or treatment with an antioxidant.
The FEBS journal (2006-07-05)
Wance J J Firdaus, Andreas Wyttenbach, Chantal Diaz-Latoud, R W Currie, André-Patrick Arrigo
ABSTRACT
We recently reported that the transient expression of polyglutamine tracts of various size in exon 1 of the huntingtin polypeptide (httEx1) generated abnormally high levels of intracellular reactive oxygen species that directly contributed to cell death. Here, we compared the protection generated by heat shock proteins to that provided by the antioxidant agent N-acetyl-L-cysteine. In cells expressing httEx1 with 72 glutamine repeats (httEx1-72Q), the overexpression of Hsp27 or Hsp70 plus Hdj-1(Hsp40) or treatment of the cells with N-acetyl-L-cysteine inhibited not only mitochondrial membrane potential disruption but also the increase in reactive oxygen species, nitric oxide and protein oxidation. However, only heat shock proteins and not N-acetyl-L-cysteine reduced the size of the inclusion bodies formed by httEx1-72Q. In cells expressing httEx1 polypeptide with 103 glutamine repeats (httEx1-103Q), heat shock proteins neither decreased oxidative damage nor reduced the size of the inclusions. In contrast, N-acetyl-L-cysteine still efficiently decreased the oxidative damage induced by httEx1-103Q polypeptide without altering the inclusions. N-Acetyl-L-cysteine was inactive with regard to proteasome inhibition, whereas heat shock proteins partially restored the caspase-like activity of this protease. These observations suggest some relationships between the presence of inclusion bodies and the oxidative damage induced by httEx1-polyQ.
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, BioXtra
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.