MilliporeSigma
  • Growth arrest-specific 1 binds to and controls the maturation and processing of the amyloid-beta precursor protein.
Growth arrest-specific 1 binds to and controls the maturation and processing of the amyloid-beta precursor protein.
Human molecular genetics (2011-03-02)
Julien Chapuis, Valérie Vingtdeux, Fabien Campagne, Peter Davies, Philippe Marambaud
ABSTRACT
Alzheimer's disease (AD), the most common neurodegenerative disorder, is characterized by cerebral deposition of amyloid-β (Aβ), a series of peptides derived from the processing of the amyloid-β precursor protein (APP). To identify new candidate genes for AD, we recently performed a transcriptome analysis to screen for genes preferentially expressed in the hippocampus and located in AD linkage regions. This strategy identified CALHM1 (calcium homeostasis modulator 1), a gene modulating AD age at onset and Aβ metabolism. Here, we focused our attention on another candidate identified using this screen, growth arrest-specific 1 (Gas1), a gene involved in the central nervous system development. We found that Gas1 formed a complex with APP and controlled APP maturation and processing. Gas1 expression inhibited APP full glycosylation and routing to the cell surface by leading to a trafficking blockade of APP between the endoplasmic reticulum and the Golgi. Gas1 expression also resulted in a robust inhibition of APP transport into multivesicular bodies, further demonstrating that Gas1 negatively regulated APP intracellular trafficking. Consequently, Gas1 overexpression led to a reduction in Aβ production, and conversely, Gas1 silencing in cells expressing endogenously Gas1 increased Aβ levels. These results suggest that Gas1 is a novel APP-interacting protein involved in the control of APP maturation and processing.
Sigma-Aldrich
Bafilomycin A1, Streptomyces griseus, Bafilomycin A1, CAS 88899-55-2, acts as a highly potent and specific inhibitor of vacuolar-type H+-ATPase (Ki = 500 pM). Blocks the fusion of autophagosome with lysosome.
Sigma-Aldrich
Enzymatic Protein Deglycosylation Kit