MilliporeSigma
  • A nonhuman primate model of early Alzheimer's disease pathologic change: Implications for disease pathogenesis.
A nonhuman primate model of early Alzheimer's disease pathologic change: Implications for disease pathogenesis.
Alzheimer's & dementia : the journal of the Alzheimer's Association (2018-11-24)
Caitlin S Latimer, Carol A Shively, C Dirk Keene, Matthew J Jorgensen, Rachel N Andrews, Thomas C Register, Thomas J Montine, Angela M Wilson, Bryan J Neth, Akiva Mintz, Joseph A Maldjian, Christopher T Whitlow, Jay R Kaplan, Suzanne Craft
ABSTRACT
Nonhuman primates may serve as excellent models of sporadic age-associated brain β-amyloid deposition and Alzheimer's disease pathologic changes. We examined whether a vervet nonhuman primate model recapitulated pathologic, physiologic, and behavioral features of early Alzheimer's disease. Nine middle-aged (mean = 11.2 years) and nine aged (mean = 21.7 years) female vervet/African green monkeys underwent cerebrospinal fluid collection, gait speed measurement, and neuroimaging before neuropathologic assessment. β-amyloid plaques were identified in all aged vervets and paired helical filament tau immunoreactivity was observed in all animals. Cerebrospinal fluid β-amyloid42 and gait speed correlated negatively with age and plaque density. Greater plaque and paired helical filament tau burden predicted reduced volumes and CMRg in several brain regions. We observed a coordinated set of relationships among neuropathologic, cerebrospinal fluid, imaging, and behavioral modalities consistent with early Alzheimer's disease. Our results support future use of the vervet model to explore disease mechanisms, biomarkers, and novel therapeutic strategies.
Product Description
Millipore
MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel - Multiplex Assay, allows quantitative multiplex detection of multiple analytes simultaneously