MilliporeSigma
  • Interphase adhesion geometry is transmitted to an internal regulator for spindle orientation via caveolin-1.
Interphase adhesion geometry is transmitted to an internal regulator for spindle orientation via caveolin-1.
Nature communications (2016-06-14)
Shigeru Matsumura, Tomoko Kojidani, Yuji Kamioka, Seiichi Uchida, Tokuko Haraguchi, Akatsuki Kimura, Fumiko Toyoshima
ABSTRACT
Despite theoretical and physical studies implying that cell-extracellular matrix adhesion geometry governs the orientation of the cell division axis, the molecular mechanisms that translate interphase adhesion geometry to the mitotic spindle orientation remain elusive. Here, we show that the cellular edge retraction during mitotic cell rounding correlates with the spindle axis. At the onset of mitotic cell rounding, caveolin-1 is targeted to the retracting cortical region at the proximal end of retraction fibres, where ganglioside GM1-enriched membrane domains with clusters of caveola-like structures are formed in an integrin and RhoA-dependent manner. Furthermore, Gαi1-LGN-NuMA, a well-known regulatory complex of spindle orientation, is targeted to the caveolin-1-enriched cortical region to guide the spindle axis towards the cellular edge retraction. We propose that retraction-induced cortical heterogeneity of caveolin-1 during mitotic cell rounding sets the spindle orientation in the context of adhesion geometry.
Sigma-Aldrich
Anti-c-Abl (Ab-3) Mouse mAb (24-21), liquid, clone 24-21, Calbiochem®