MilliporeSigma
  • Super-resolution imaging reveals α-synuclein seeded aggregation in SH-SY5Y cells.
Super-resolution imaging reveals α-synuclein seeded aggregation in SH-SY5Y cells.
Communications biology (2021-05-23)
Jason C Sang, Eric Hidari, Georg Meisl, Rohan T Ranasinghe, Maria Grazia Spillantini, David Klenerman
ABSTRACT
Aggregation of α-synuclein (α-syn) is closely linked to Parkinson's disease (PD) and the related synucleinopathies. Aggregates spread through the brain during the progression of PD, but the mechanism by which this occurs is still not known. One possibility is a self-propagating, templated-seeding mechanism, but this cannot be established without quantitative information about the efficiencies and rates of the key steps in the cellular process. To address this issue, we imaged the uptake and seeding of unlabeled exogenous α-syn fibrils by SH-SY5Y cells and the resulting secreted aggregates, using super-resolution microscopy. Externally-applied fibrils very inefficiently induced self-assembly of endogenous α-syn in a process accelerated by the proteasome. Seeding resulted in the increased secretion of nanoscopic aggregates (mean 35 nm diameter), of both α-syn and Aβ. Our results suggest that cells respond to seed-induced disruption of protein homeostasis predominantly by secreting nanoscopic aggregates; this mechanism may therefore be an important protective response by cells to protein aggregation.
Sigma-Aldrich
NS-398, ≥98% (HPLC), solid
Sigma-Aldrich
Tumor Necrosis Factor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
BioPORTER® QuikEase Protein Delivery Kit, for intracellular delivery
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor