MilliporeSigma
  • Impacts of the SOAT1 genetic variants and protein expression on HBV-related hepatocellular carcinoma.
Impacts of the SOAT1 genetic variants and protein expression on HBV-related hepatocellular carcinoma.
BMC cancer (2021-05-28)
Yulong Chen, Xunjun Yang, Yao Chen, Guorong Chen, Cheryl A Winkler, Ping An, Jianxin Lyu
ABSTRACT
Hepatitis B virus (HBV)-related hepatocellular carcinoma (HCC) remains a major public health problem and its pathogenesis remains unresolved. A recent proteomics study discovered a lipid enzyme Sterol O-acyltransferase (SOAT1) involvement in the progression of HCC. We aimed to explore the association between SOAT1 genetic variation and HCC. We genotyped three exonic SOAT1 variants (rs10753191, V323V; rs3753526, L475L; rs13306731, Q526R) tagging most variations in the gene, in 221 HCC patients and 229 healthy individuals, to assess the impact of SOAT1 gene variation on risk of HCC occurrence. We further conducted immunohistochemistry to compare SOAT1 protein expression levels in 42 paired tumor and adjacent non-tumor tissues. We found that rs10753191 (Odds ratio (OR) = 0.58, P = 0.04) and a haplotype TGA (OR = 0.40, P = 0.01) were associated with reduced HCC risk after adjusting for lipid levels. In the immunohistochemistry experiment, we found that the protein expression of SOAT1 was significantly increased in the tumor compared with adjacent tissue (P < 0.001). This study revealed for the first time SOAT1 genetic variation that associates with host susceptibility to HCC occurrence. Our results suggest a role of SOAT1 in the HCC development, which warrants further elucidation.
Sigma-Aldrich
Anti-Cholesterol acyltransferase 1 Antibody, from rabbit, purified by affinity chromatography