MilliporeSigma
  • Water-soluble allyl sulfones for dual site-specific labelling of proteins and cyclic peptides.
Water-soluble allyl sulfones for dual site-specific labelling of proteins and cyclic peptides.
Chemical science (2016-05-01)
Tao Wang, Andreas Riegger, Markus Lamla, Sebastian Wiese, Patrick Oeckl, Markus Otto, Yuzhou Wu, Stephan Fischer, Holger Barth, Seah Ling Kuan, Tanja Weil
ABSTRACT
Water-soluble allyl sulfones provide convenient site-specific disulfide rebridging of native proteins and cyclic peptides. The site-selective functionalization of (a) the peptide hormone somatostatin, (b) the interchain disulfide of bovine insulin and (c) functionalization of the proteins GFP and lysozyme with allyl sulfones proceeds in aqueous solution. Allyl sulfones offer three functionalizable sites that react with thiol containing molecules in a step-wise fashion. Dual labeling of proteins and cyclic peptides is achieved i.e. the attachment of a chromophore and an affinity tag in a single reaction step, which is of great significance for the construction of precise multifunctional peptide and protein conjugates.
Sigma-Aldrich
Lysozyme Activity Kit, sufficient for 100 tests