MilliporeSigma
  • Genome-scale CRISPR-Cas9 screen of Wnt/β-catenin signaling identifies therapeutic targets for colorectal cancer.
Genome-scale CRISPR-Cas9 screen of Wnt/β-catenin signaling identifies therapeutic targets for colorectal cancer.
Science advances (2021-06-18)
Chunhua Wan, Sylvia Mahara, Claire Sun, Anh Doan, Hui Kheng Chua, Dakang Xu, Jia Bian, Yue Li, Danxi Zhu, Dhanya Sooraj, Tomasz Cierpicki, Jolanta Grembecka, Ron Firestein
ABSTRACT
Aberrant activation of Wnt/β-catenin pathway is a key driver of colorectal cancer (CRC) growth and of great therapeutic importance. In this study, we performed comprehensive CRISPR screens to interrogate the regulatory network of Wnt/β-catenin signaling in CRC cells. We found marked discrepancies between the artificial TOP reporter activity and β-catenin-mediated endogenous transcription and redundant roles of T cell factor/lymphoid enhancer factor transcription factors in transducing β-catenin signaling. Compiled functional genomic screens and network analysis revealed unique epigenetic regulators of β-catenin transcriptional output, including the histone lysine methyltransferase 2A oncoprotein (KMT2A/Mll1). Using an integrative epigenomic and transcriptional profiling approach, we show that KMT2A loss diminishes the binding of β-catenin to consensus DNA motifs and the transcription of β-catenin targets in CRC. These results suggest that KMT2A may be a promising target for CRCs and highlight the broader potential for exploiting epigenetic modulation as a therapeutic strategy for β-catenin-driven malignancies.
Sigma-Aldrich
GenElute HP 96-Well Plasmid Miniprep Kit, sufficient for 4 96-well plate purifications
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture