MilliporeSigma
  • Combined fluorescent and interferometric detection of protein on a BioCD.
Combined fluorescent and interferometric detection of protein on a BioCD.
Applied optics (2008-05-22)
Xuefeng Wang, Ming Zhao, D D Nolte
ABSTRACT
We perform simultaneous interferometric and fluorescent detection of molecular protein layers on a BioCD. The 488 nm excitation wavelength of fluorescein also provides the interferometric detection channel that operates in a common-path in-line configuration in the condition of phase quadrature set by a thermal oxide on silicon. The simultaneous acquisition of both channels enables a direct correlation between bound mass and fluorescent surface density, which we compare in forward- and reverse-phase immunoassays. Scaling mass sensitivities for immunoassays measured in the interferometric and fluorescent channels are 15 pg/mm and 1.5 pg/mm, respectively, when applied to gel-printed periodic antibody patterns detected in the frequency domain from the spinning disc. These sensitivities are limited by the inhomogeneities of the print. While fluorescence is subject to bleaching, the interferometry signal is robust under long-term laser illumination.
Sigma-Aldrich
Anti-Avidin–FITC antibody, Mouse monoclonal, clone WC19.10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Goat IgG (whole molecule) antibody produced in donkey, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule) antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) antibody produced in goat, affinity isolated antibody, lyophilized powder