MilliporeSigma
  • Predictive Biomarkers for Endocrine Therapy: Retrospective Study in Tamoxifen and Exemestane Adjuvant Multinational (TEAM) Trial.
Predictive Biomarkers for Endocrine Therapy: Retrospective Study in Tamoxifen and Exemestane Adjuvant Multinational (TEAM) Trial.
Journal of the National Cancer Institute (2018-06-20)
Antonia K Roseweir, Lindsay Bennett, Ashley Dickson, Kelvin Cheng, Mary-Anne Quintayo, Jane Bayani, Donald C McMillan, Paul G Horgan, Cornelis J H van de Velde, Caroline Seynaeve, Annette Hasenburg, Dirk G Kieback, Christos Markopoulos, Luc Y Dirix, Daniel W Rea, Elizabeth A Mallon, John M S Bartlett, Joanne Edwards
ABSTRACT
Aromatase inhibitors improve disease-free survival compared with tamoxifen in postmenopausal women with hormone receptor-positive breast cancer. The Tamoxifen and Exemestane Adjuvant Multinational (TEAM) trial compared exemestane monotherapy with sequential therapy of tamoxifen followed by exemestane. The trial failed to show a statistically significant difference between treatment arms. A robust translational program was established to investigate predictive biomarkers. A tissue microarray was retrospectively constructed using a subset of patient tissues (n = 4631) from the TEAM trial (n = 9766). Immunohistochemistry was performed for biomarkers, classed into three groups: MAPK pathway, NF-kappa B pathway, and estrogen receptor (ER) phosphorylation. Expression was analyzed for association with relapse-free survival (RFS) at 2.5 and 10 years and treatment regimen using Kaplan-Meier curves and log-rank analysis. All statistical tests were two-sided. In univariate analysis, ER167 (hazard ratio [HR] = 0.71, 95% confidence interval [CI] = 0.59 to 0.85, P < .001), IKKα (HR = 0.74, 95% CI = 0.60 to 0.92, P = .005), Raf-1338 (HR = 0.64, 95% CI = 0.52 to 0.80, P < .001), and p44/42 MAPK202/204 (HR = 0.77, 95% CI = 0.64 to 0.92, P = .004) were statistically significantly associated with improved RFS at 10 years in patients receiving sequential therapy. Associations were strengthened when IKKα, Raf-1338, and ER167 were combined into a cumulative prognostic score (HR = 0.64, 95% CI = 0.52 to 0.77, P < .001). Patients with an all negative IKKα, Raf-1338, and ER167 score favored exemestane monotherapy (odds ratio = 0.56, 95% CI = 0.35 to 0.90). In multivariable analysis, the IKKα, Raf-1338, and ER167 score (P = .001) was an independent prognostic factor for RFS at 10 years in patients receiving sequential therapy. The IKKα, Raf-1338, and ER167 score is an independent predictive biomarker for lower recurrence on sequential therapy. Negative expression may further offer predictive value for exemestane monotherapy.
Sigma-Aldrich
Anti-phospho-Raf-1 (Ser338) Antibody, Upstate®, from mouse